siRNA Details
| Gene Symbol: MED10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032286.2 | NP_115662.2 | 2 | 222 | ||
| GenBank mRNA | AY277599.1 | - | 2 | 179 | ||
| AY453397.1 | - | 2 | 220 | |||
| AY598325.1 | - | 2 | 179 | |||
| BC003353.1 | - | 2 | 222 | 2899986 | ||
| BI597082.1 | - | 2 | 211 | |||

MED10
Genomic Map