siRNA Details
| Gene Symbol: SLC37A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287498.1 | NP_001274427.1 | 6 | 618 | ||
| NM_032295.3 | NP_115671.1 | 6 | 618 | |||
| NM_207113.2 | NP_996996.1 | 6 | 618 | |||
| XM_011516626.2 | XP_011514928.1 | 8 | 1115 | |||
| XM_011516627.2 | XP_011514929.1 | 8 | 1114 | |||
| XM_017012712.1 | XP_016868201.1 | 8 | 1091 | |||
| XM_017012713.1 | XP_016868202.1 | 8 | 1110 | |||
| XM_017012715.1 | XP_016868204.1 | 8 | 1134 | |||
| XM_017012716.1 | XP_016868205.1 | 8 | 1138 | |||
| XM_017012717.1 | XP_016868206.1 | 8 | 1110 | |||
| XM_017012718.1 | XP_016868207.1 | 8 | 1109 | |||
| GenBank mRNA | AK074823.1 | - | 6 | 579 | ||
| AK296543.1 | - | 5 | 488 | |||
| AK299276.1 | - | 6 | 562 | |||
| AL136583.1 | - | 6 | 525 | |||
| BC028380.1 | - | 6 | 568 | 4811814 | ||
| BC046567.1 | - | 6 | 570 | 5246993 | ||

SLC37A3
Genomic Map