siRNA Details
| Gene Symbol: FBXW9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032301.2 | NP_115677.2 | 3 | 720 | ||
| XM_005260096.4 | XP_005260153.1 | 2 | 839 | |||
| GenBank mRNA | AK123037.1 | - | 3 | 719 | ||
| AK302744.1 | - | 2 | 558 | |||
| AY736034.1 | - | 3 | 630 | |||
| BC004290.1 | - | 3 | 697 | |||

FBXW9
Genomic Map