siRNA Details
| Gene Symbol: FBXW9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032301.2 | NP_115677.2 | 9 | 1320 | ||
| XM_005260096.4 | XP_005260153.1 | 8 | 1439 | |||
| GenBank mRNA | AK123037.1 | - | 9 | 1319 | ||
| AK302744.1 | - | 7 | 1045 | |||
| AY736034.1 | - | 9 | 1320 | |||
| BC004290.1 | - | 9 | 1297 | |||

FBXW9
Genomic Map