siRNA Details
| Gene Symbol: ALKBH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032306.3 | NP_115682.1 | 1 | 570 | ||
| XM_017027355.1 | XP_016882844.1 | 1 | 206 | |||
| GenBank mRNA | AK223336.1 | - | 1 | 193 | ||
| AK311956.1 | - | 1 | 209 | |||
| AY358858.1 | - | 1 | 570 | |||
| AY427650.2 | - | 1 | 570 | |||
| BC004393.1 | - | 1 | 225 | 3635540 | ||
| BX334356.2 | - | 1 | 487 | |||

ALKBH7
Genomic Map