siRNA Details
| Gene Symbol: C9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317997.1 | NP_001304926.1 | 4 | 560 | ||
| NM_032307.4 | NP_115683.3 | 3 | 958 | |||
| GenBank mRNA | AK090882.1 | - | 4 | 560 | ||
| BC004407.1 | - | 3 | 933 | |||
| BC137461.1 | - | 3 | 835 | 9021461 | ||
| BC137462.1 | - | 3 | 835 | 9021462 | ||

C9orf64
Genomic Map