siRNA Details
| Gene Symbol: PRADC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032319.1 | NP_115695.1 | 2 | 226 | ||
| GenBank mRNA | AK312077.1 | - | 2 | 234 | ||
| AY358605.1 | - | 2 | 217 | |||
| BC005069.1 | - | 2 | 226 | 3530601 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRADC1| Gene Symbol: PRADC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032319.1 | NP_115695.1 | 2 | 226 | ||
| GenBank mRNA | AK312077.1 | - | 2 | 234 | ||
| AY358605.1 | - | 2 | 217 | |||
| BC005069.1 | - | 2 | 226 | 3530601 | ||