siRNA Details
| Gene Symbol: C2orf88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042519.1 | NP_001035984.1 | 2 | 510 | ||
| NM_001042520.1 | NP_001035985.1 | 2 | 466 | |||
| NM_001042521.1 | NP_001035986.1 | 3 | 478 | |||
| NM_032321.2 | NP_115697.2 | 2 | 437 | |||
| XM_005246905.1 | XP_005246962.1 | 4 | 659 | |||
| XM_006712795.2 | XP_006712858.1 | 2 | 344 | |||
| XM_011511982.1 | XP_011510284.1 | 5 | 746 | |||
| XM_011511983.1 | XP_011510285.1 | 5 | 811 | |||
| XM_011511984.2 | XP_011510286.1 | 5 | 602 | |||
| XM_011511985.2 | XP_011510287.1 | 4 | 518 | |||
| XM_011511986.2 | XP_011510288.1 | 3 | 682 | |||
| XM_017005096.1 | XP_016860585.1 | 2 | 526 | |||
| GenBank mRNA | AK075168.1 | - | 2 | 1192 | ||
| AK313395.1 | - | 2 | 406 | |||
| BC005083.1 | - | 2 | 437 | 3632431 | ||
| BU567236.1 | - | 3 | 486 | |||
| BX648070.1 | - | 1 | 15 | |||
| DA247977.1 | - | 2 | 510 | |||
| DA389238.1 | - | 2 | 467 | |||

C2orf88
Genomic Map