siRNA Details
| Gene Symbol: TMEM175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032326.3 | NP_115702.1 | 3, 4 | 380 | ||
| XM_005272301.2 | XP_005272358.1 | 3, 4 | 355 | |||
| XM_005272304.1 | XP_005272361.1 | 3, 4 | 310 | |||
| XM_005272307.4 | XP_005272364.1 | 3, 4 | 306 | |||
| XM_017008701.1 | XP_016864190.1 | 4, 5 | 457 | |||
| GenBank mRNA | AK309132.1 | - | 3, 4 | 306 | ||
| BC005158.2 | - | 3, 4 | 293 | 3505819 | ||

TMEM175
Genomic Map