siRNA Details
| Gene Symbol: WDR83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099737.2 | NP_001093207.1 | 6 | 694 | ||
| NM_032332.3 | NP_115708.1 | 4 | 437 | |||
| NR_029375.1 | 6 | 677 | ||||
| GenBank mRNA | AK074525.1 | - | 6 | 667 | ||
| AK223195.1 | - | 4 | 387 | |||
| AK314164.1 | - | 6 | 671 | |||
| BC005870.2 | - | 4 | 370 | 2961616 | ||
| CB143206.1 | - | 4 | 437 | |||

WDR83
Genomic Map