siRNA Details
| Gene Symbol: MIEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032339.4 | NP_115715.3 | 4 | 555 | ||
| XM_005257736.3 | XP_005257793.1 | 3 | 613 | |||
| GenBank mRNA | AL554581.3 | - | 4 | 777 | ||
| AY508814.1 | - | 4 | 555 | |||
| BC006006.1 | - | 4 | 494 | 4283597 | ||
| BC106016.1 | - | 2 | 702 | |||
| BX646129.1 | - | 4 | 528 | |||

MIEN1
Genomic Map