siRNA Details
| Gene Symbol: UQCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032340.3 | NP_115716.1 | 2 | 254 | ||
| GenBank mRNA | AF086234.1 | - | 2 | 176 | ||
| AK311835.1 | - | 2 | 203 | |||
| AY568085.1 | - | 2 | 230 | |||
| BC006007.1 | - | 2 | 230 | 4284544 | ||
| BI604191.1 | - | 2 | 204 | |||
| DA123098.1 | - | 2 | 254 | |||
| EF139858.1 | - | 4 | 504 | |||

UQCC2
Genomic Map