siRNA Details
| Gene Symbol: PYM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143853.1 | NP_001137325.1 | 3 | 159 | ||
| NM_032345.2 | NP_115721.1 | 3 | 298 | |||
| GenBank mRNA | AJ459406.1 | - | 3 | 146 | ||
| AK096922.1 | - | 3 | 146 | |||
| BC006135.1 | - | 3 | 298 | 3640407 | ||
| BC014976.1 | - | 3 | 267 | 4854916 | ||
| DB466388.1 | - | 3 | 159 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001170869, NM_030100, NM_001253704, NM_001253705, AJ459407, and others.

PYM1
Genomic Map