siRNA Details
| Gene Symbol: ZNF397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135178.2 | NP_001128650.1 | 4 | 793 | ||
| NM_032347.2 | NP_115723.1 | 4 | 793 | |||
| XM_006722558.2 | XP_006722621.1 | 4 | 846 | |||
| XM_011526229.2 | XP_011524531.1 | 5 | 935 | |||
| XM_011526230.2 | XP_011524532.1 | 4 | 1137 | |||
| XM_011526231.2 | XP_011524533.1 | 4 | 734 | |||
| XM_017026042.1 | XP_016881531.1 | 4 | 789 | |||
| GenBank mRNA | AF533250.2 | - | 4 | 786 | ||
| AK311533.1 | - | 3 | 278 | |||
| BC006172.1 | - | 4 | 768 | 4053312 | ||

ZNF397
Genomic Map