siRNA Details
| Gene Symbol: C7orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134395.1 | NP_001127867.1 | 3 | 274 | ||
| NM_001134396.1 | NP_001127868.1 | 3 | 255 | |||
| NM_001318252.1 | NP_001305181.1 | 3 | 1158 | |||
| NM_032350.5 | NP_115726.1 | 3 | 284 | |||
| NR_134537.1 | 2 | 260 | ||||
| XM_006715793.2 | XP_006715856.1 | 2 | 898 | |||
| XM_011515580.2 | XP_011513882.1 | 2 | 898 | |||
| XM_011515581.2 | XP_011513883.1 | 3 | 318 | |||
| XM_011515582.2 | XP_011513884.1 | 3 | 269 | |||
| XM_011515583.2 | XP_011513885.1 | 3 | 293 | |||
| XM_011515584.2 | XP_011513886.1 | 3 | 974 | |||
| XM_017012719.1 | XP_016868208.1 | 2 | 898 | |||
| XM_017012720.1 | XP_016868209.1 | 3 | 278 | |||
| XM_017012721.1 | XP_016868210.1 | 4 | 1197 | |||
| XM_017012722.1 | XP_016868211.1 | 3 | 317 | |||
| GenBank mRNA | AF495721.1 | - | 3 | 1114 | ||
| BC006224.2 | - | 3 | 284 | 3941780 | ||
| BF129057.1 | - | 260 | ||||
| BM762287.1 | - | 2 | 204 | |||
| BM763235.1 | - | 3 | 258 | |||
| CN261680.1 | - | 3 | 301 | |||

C7orf50
Genomic Map