siRNA Details
| Gene Symbol: HVCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040107.1 | NP_001035196.1 | 5 | 582 | ||
| NM_001256413.1 | NP_001243342.1 | 4 | 542 | |||
| NM_032369.3 | NP_115745.2 | 5 | 573 | |||
| XM_005253948.2 | XP_005254005.1 | 5 | 660 | |||
| XM_011538838.1 | XP_011537140.1 | 6 | 998 | |||
| XM_011538839.2 | XP_011537141.1 | 6 | 732 | |||
| XM_011538840.2 | XP_011537142.1 | 6 | 764 | |||
| XM_011538841.2 | XP_011537143.1 | 6 | 742 | |||
| XM_011538842.2 | XP_011537144.1 | 6 | 722 | |||
| XM_011538844.2 | XP_011537146.1 | 6 | 737 | |||
| XM_011538845.1 | XP_011537147.1 | 5 | 617 | |||
| XM_011538846.2 | XP_011537148.1 | 6 | 691 | |||
| XM_011538847.1 | XP_011537149.1 | 5 | 592 | |||
| XM_017020026.1 | XP_016875515.1 | 6 | 706 | |||
| XM_017020027.1 | XP_016875516.1 | 5 | 591 | |||
| GenBank mRNA | AK293543.1 | - | 4 | 508 | ||
| AY359054.1 | - | 5 | 601 | |||
| BC007277.1 | - | 5 | 1025 | 3343102 | ||
| BC009731.1 | - | 5 | 524 | 3877292 | ||
| BC032672.2 | - | 4 | 1146 | 5577070 | ||
| BP369986.1 | - | 5 | 455 | |||
| GU211002.1 | - | 3 | 392 | |||

HVCN1
Genomic Map