siRNA Details
| Gene Symbol: ZNF414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146175.1 | NP_001139647.1 | 4, 5 | 642 | ||
| NM_032370.2 | NP_115746.2 | 4, 5 | 642 | |||
| GenBank mRNA | AK074191.1 | - | 4, 5 | 638 | ||
| AK225034.1 | - | 4, 5 | 638 | |||
| BC007310.1 | - | 4, 5 | 626 | 3353224 | ||
| BC071937.1 | - | 4, 5 | 621 | 5089259 | ||

ZNF414
Genomic Map