siRNA Details
| Gene Symbol: PCGF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256549.1 | NP_001243478.1 | 4 | 581 | ||
| NM_001257101.1 | NP_001244030.1 | 4 | 474 | |||
| NM_032373.4 | NP_115749.2 | 4 | 639 | |||
| XM_006718030.1 | XP_006718093.1 | 3 | 570 | |||
| XM_011540271.2 | XP_011538573.1 | 4 | 640 | |||
| XM_011540272.2 | XP_011538574.1 | 4 | 671 | |||
| XM_017016776.1 | XP_016872265.1 | 4 | 586 | |||
| XM_017016777.1 | XP_016872266.1 | 4 | 585 | |||
| XM_017016778.1 | XP_016872267.1 | 3 | 570 | |||
| GenBank mRNA | AK303014.1 | - | 4 | 582 | ||
| AL832003.2 | - | 2 | 78 | |||
| AL832496.1 | - | 4 | 455 | |||
| BC051845.1 | - | 4 | 449 | 5199133 | ||
| BI464238.1 | - | 4 | 645 | |||

PCGF5
Genomic Map