siRNA Details
| Gene Symbol: PRAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032391.2 | NP_115767.1 | 2 | 262 | ||
| GenBank mRNA | AF331165.1 | - | 2 | 262 | ||
| BC030950.1 | - | 2 | 260 | 4739311 | ||
| BU535394.1 | - | 2 | 265 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRAC1| Gene Symbol: PRAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032391.2 | NP_115767.1 | 2 | 262 | ||
| GenBank mRNA | AF331165.1 | - | 2 | 262 | ||
| BC030950.1 | - | 2 | 260 | 4739311 | ||
| BU535394.1 | - | 2 | 265 | |||