siRNA Details
| Gene Symbol: C15orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032413.3 | NP_115789.1 | 2, 3 | 386 | ||
| NM_197955.2 | NP_922946.1 | 3, 4 | 272 | |||
| GenBank mRNA | AB026707.1 | - | 3, 4 | 166 | ||
| AF228422.1 | - | 2, 3 | 307 | |||
| AK026298.1 | - | 2, 3 | 304 | |||
| BC021173.2 | - | 3, 4 | 192 | 5269504 | ||
| BM312626.1 | - | 3, 4 | 236 | |||
| CA865860.1 | - | 3, 4 | 278 | |||
| DB447060.1 | - | 3, 4 | 274 | |||

C15orf48
Genomic Map