siRNA Details
| Gene Symbol: ADGRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083909.2 | NP_001077378.1 | 4 | 563 | ||
| XM_005252695.1 | XP_005252752.1 | 13 | 2386 | |||
| XM_011540273.1 | XP_011538575.1 | 1 | 40 | |||
| XM_017016779.1 | XP_016872268.1 | 3 | 423 | |||
| GenBank mRNA | AY181241.1 | - | 13 | 2386 | ||
| BG818167.1 | - | 3 | 298 | |||
| EF536972.1 | - | 3 | 352 | |||
More Information
Set Membership: |
|

ADGRA1
Genomic Map