siRNA Details
Gene Symbol: ADGRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083909.2 | NP_001077378.1 | 7 | 2910 | ||
NM_001291085.1 | NP_001278014.1 | 4 | 2411 | |||
XM_005252695.1 | XP_005252752.1 | 16 | 4730 | |||
XM_011540273.1 | XP_011538575.1 | 3 | 2241 | |||
GenBank mRNA | AB058731.1 | - | 4 | 2511 | ||
AK090982.1 | - | 1 | 1298 | |||
AK094675.1 | - | 1 | 738 | |||
AK124728.1 | - | 5 | 951 | |||
BC044247.1 | - | 1 | 698 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
