siRNA Details
| Gene Symbol: FRMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032428.1 | NP_115804.1 | 3 | 176 | ||
| XM_017029901.1 | XP_016885390.1 | 1 | 397 | |||
| XM_017029902.1 | XP_016885391.1 | 1 | 397 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FRMPD3| Gene Symbol: FRMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032428.1 | NP_115804.1 | 3 | 176 | ||
| XM_017029901.1 | XP_016885390.1 | 1 | 397 | |||
| XM_017029902.1 | XP_016885391.1 | 1 | 397 | |||