siRNA Details
| Gene Symbol: BRSK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032430.1 | NP_115806.1 | 1, 2 | 416 | ||
| XM_005259327.3 | XP_005259384.1 | 1, 2 | 481 | |||
| GenBank mRNA | AF479826.1 | - | 3, 4 | 314 | ||
| AF479827.1 | - | 1, 2 | 416 | |||
| AL834275.1 | - | 1, 2 | 39 | |||
| AY458602.1 | - | 1, 2 | 188 | |||
| AY505124.1 | - | 1, 2 | 139 | |||
| HQ830199.1 | - | 1, 2 | 161 | |||

BRSK1
Genomic Map