siRNA Details
| Gene Symbol: BRSK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032430.1 | NP_115806.1 | 9 | 1125 | ||
| XM_005259327.3 | XP_005259384.1 | 9 | 1194 | |||
| XM_011527395.2 | XP_011525697.2 | 7 | 559 | |||
| GenBank mRNA | AB058714.1 | - | 8 | 660 | ||
| AF479826.1 | - | 11 | 1023 | |||
| AF479827.1 | - | 9 | 1125 | |||
| AK124999.1 | - | 1 | 94 | |||
| AL834275.1 | - | 9 | 748 | |||
| AY458602.1 | - | 9 | 897 | |||
| AY505124.1 | - | 9 | 848 | |||
| HQ830199.1 | - | 9 | 870 | |||

BRSK1
Genomic Map