siRNA Details
| Gene Symbol: SPIRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032451.1 | NP_115827.1 | 11 | 1724 | ||
| XM_011523379.1 | XP_011521681.1 | 11 | 2490 | |||
| XM_011523380.2 | XP_011521682.1 | 12 | 1746 | |||
| XM_011523381.1 | XP_011521683.1 | 12 | 2570 | |||
| XM_011523384.1 | XP_011521686.1 | 11 | 1748 | |||
| XM_017023779.1 | XP_016879268.1 | 11 | 1736 | |||
| XM_017023780.1 | XP_016879269.1 | 11 | 1901 | |||
| XM_017023781.1 | XP_016879270.1 | 10 | 1373 | |||
| GenBank mRNA | AB058735.1 | - | 9 | 1822 | ||
| AJ422077.1 | - | 11 | 1714 | |||
| AK094918.1 | - | 4 | 541 | |||
| AK130685.1 | - | 1 | 1606 | |||
| AL834408.1 | - | 11 | 1518 | |||
| BC063706.1 | - | 3 | 337 | |||
| BC111030.1 | - | 11 | 1724 | 5244492 | ||
| BC139732.1 | - | 9 | 1342 | 40075864 | ||

SPIRE2
Genomic Map