siRNA Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032453.1 | NP_115829.1 | 5 | 1558 | ||
| XM_005259328.4 | XP_005259385.1 | 5 | 1650 | |||
| XM_005259329.3 | XP_005259386.1 | 4 | 1564 | |||
| XM_017027380.1 | XP_016882869.1 | 3 | 1539 | |||
| XM_017027381.1 | XP_016882870.1 | 4 | 1550 | |||
| GenBank mRNA | AB058732.1 | - | 2 | 1291 | ||
| AK091585.1 | - | 4 | 1453 | |||
| AK301135.1 | - | 5 | 1554 | |||

ZNF527
Genomic Map