siRNA Details
| Gene Symbol: ACRBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032489.2 | NP_115878.2 | 2 | 166 | ||
| GenBank mRNA | AB051833.1 | - | 2 | 166 | ||
| AK302290.1 | - | 2 | 163 | |||
| AK311572.1 | - | 2 | 163 | |||
| AK312782.1 | - | 2 | 184 | |||
| BC033010.2 | - | 2 | 163 | 4822765 | ||

ACRBP
Genomic Map