siRNA Details
| Gene Symbol: GON7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032490.4 | NP_115879.2 | 2 | 821 | ||
| GenBank mRNA | BC021733.2 | - | 2 | 801 | 4796574 | |
| BC065216.1 | - | 2 | 723 | |||
| BC107429.1 | - | 2 | 789 | 4809642 | ||
| BC109390.1 | - | 2 | 732 | |||

GON7
Genomic Map