siRNA Details
| Gene Symbol: ZC3H8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032494.2 | NP_115883.2 | 1, 2 | 188 | ||
| GenBank mRNA | AF334161.1 | - | 1, 2 | 174 | ||
| AK315329.1 | - | 1, 2 | 142 | |||
| BC032001.1 | - | 1, 2 | 126 | 4749794 | ||
| BG773518.1 | - | 1, 2 | 217 | |||

ZC3H8
Genomic Map