siRNA Details
| Gene Symbol: ACSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252675.1 | NP_001239604.1 | 2 | 484 | ||
| NM_001252677.1 | NP_001239606.1 | 2 | 484 | |||
| NM_032501.3 | NP_115890.2 | 2 | 484 | |||
| XM_006723659.1 | XP_006723722.1 | 2 | 445 | |||
| XM_011529388.1 | XP_011527690.1 | 2 | 445 | |||
| GenBank mRNA | AK094414.1 | - | 2 | 1283 | ||
| AK127566.1 | - | 2 | 430 | |||
| AK314440.1 | - | 2 | 407 | |||
| AL832939.1 | - | 2 | 168 | |||
| BC039261.1 | - | 2 | 443 | 5298849 | ||
| BC044588.1 | - | 2 | 409 | 5260496 | ||
| BC055008.1 | - | 2 | 409 | 4838976 | ||
| BC114612.1 | - | 2 | 275 | |||
| BM150326.1 | - | 2 | 484 | |||

ACSS1
Genomic Map