siRNA Details
| Gene Symbol: KBTBD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032505.2 | NP_115894.2 | 2, 3 | 276 | ||
| GenBank mRNA | AF385438.1 | - | 1, 2 | 209 | ||
| AK096640.1 | - | 2, 3 | 272 | |||
| BC117487.1 | - | 1, 2 | 173 | 40126038 | ||
| DA246470.1 | - | 2, 3 | 272 | |||
| DB065239.1 | - | 2, 3 | 277 | |||

KBTBD8
Genomic Map