siRNA Details
| Gene Symbol: ZNF594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032530.1 | NP_115919.1 | 2 | 307 | ||
| XM_005256827.2 | XP_005256884.1 | 2 | 620 | |||
| GenBank mRNA | AB058774.2 | - | 1 | 292 | ||
| AK131271.1 | - | 2 | 307 | |||
| BC136604.1 | - | 1 | 285 | 9020594 | ||
| BX472376.1 | - | 2 | 303 | |||
| DA448705.1 | - | 2 | 307 | |||

ZNF594
Genomic Map