siRNA Details
| Gene Symbol: FNDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032532.2 | NP_115921.2 | 16 | 5020 | ||
| XM_011536190.2 | XP_011534492.1 | 15 | 4943 | |||
| XM_011536191.2 | XP_011534493.1 | 13 | 4661 | |||
| GenBank mRNA | AB058769.2 | - | 14 | 4488 | ||
| AJ586132.1 | - | 6 | 921 | |||
| AK127635.1 | - | 6 | 1010 | |||
| AL832410.1 | - | 6 | 1219 | |||
| BC146783.1 | - | 14 | 4488 | 9007239 | ||
| BC150607.1 | - | 15 | 4695 | 9056977 | ||
| DQ009660.1 | - | 16 | 5002 | |||

FNDC1
Genomic Map