siRNA Details
| Gene Symbol: DGKQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001347.3 | NP_001338.2 | 16 | 1915 | ||
| XM_011513411.1 | XP_011511713.1 | 16 | 1900 | |||
| XM_011513412.1 | XP_011511714.1 | 16 | 1900 | |||
| XM_017007814.1 | XP_016863303.1 | 17 | 1914 | |||
| GenBank mRNA | AB209503.1 | - | 16 | 1666 | ||
| BC063801.1 | - | 16 | 1923 | 6526640 | ||
| GU727647.1 | - | 16 | 1853 | |||
| L38707.1 | - | 16 | 1875 | |||

DGKQ
Genomic Map