siRNA Details
| Gene Symbol: AFAP1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF474151.1 | - | 7 | 770 | ||
| AK123108.1 | - | 8 | 814 | |||
| AK291712.1 | - | 7 | 768 | |||
| AK294969.1 | - | 8 | 929 | |||
| BC024314.1 | - | 7 | 1022 | 3900921 | ||
| BC033212.1 | - | 7 | 751 | 4909140 | ||

AFAP1L2
Genomic Map