siRNA Details
| Gene Symbol: RETNLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032579.2 | NP_115968.1 | 1 | 189 | ||
| GenBank mRNA | AF290873.1 | - | 1 | 191 | ||
| AF323084.1 | - | 1 | 198 | |||
| AY358655.1 | - | 1 | 190 | |||
| BC069318.1 | - | 1 | 162 | 7262095 | ||
| BC113502.1 | - | 1 | 123 | 8322554 | ||
| BC113528.1 | - | 1 | 123 | 8322580 | ||

RETNLB
Genomic Map