siRNA Details
| Gene Symbol: CARD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032587.3 | NP_115976.2 | 3 | 2638 | ||
| XM_017009989.1 | XP_016865478.1 | 2 | 2191 | |||
| XM_017009990.1 | XP_016865479.1 | 4 | 2872 | |||
| GenBank mRNA | AB209844.1 | - | 3 | 2458 | ||
| AF356193.1 | - | 3 | 2762 | |||
| BC093825.1 | - | 3 | 2585 | 7939670 | ||

CARD6
Genomic Map