siRNA Details
| Gene Symbol: TRIM63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032588.3 | NP_115977.2 | 1 | 225 | ||
| XM_017002559.1 | XP_016858048.1 | 1 | 225 | |||
| GenBank mRNA | AF353673.1 | - | 1 | 204 | ||
| AF361946.1 | - | 1 | 1029 | |||
| AJ276484.1 | - | 1 | 208 | |||
| AK056942.1 | - | 1 | 221 | |||
| AK297820.1 | - | 1 | 221 | |||
| BC080529.1 | - | 1 | 203 | 4862321 | ||
| BT007373.1 | - | 1 | 85 | |||
| DA900348.1 | - | 1 | 225 | |||

TRIM63
Genomic Map