siRNA Details
| Gene Symbol: SPATA22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170695.1 | NP_001164166.1 | 8 | 1013 | ||
| NM_001170696.1 | NP_001164167.1 | 7 | 884 | |||
| NM_001170697.1 | NP_001164168.1 | 8 | 1019 | |||
| NM_001170698.1 | NP_001164169.1 | 8 | 1034 | |||
| NM_001321336.1 | NP_001308265.1 | 8 | 1303 | |||
| NM_001321337.1 | NP_001308266.1 | 8 | 1351 | |||
| NM_032598.4 | NP_115987.2 | 8 | 1075 | |||
| GenBank mRNA | AF367472.1 | - | 8 | 967 | ||
| AK295299.1 | - | 8 | 1303 | |||
| AY032684.1 | - | 7 | 833 | |||
| AY035867.1 | - | 8 | 1016 | |||
| AY035868.1 | - | 8 | 1075 | |||
| BC029483.1 | - | 8 | 960 | 5271286 | ||

SPATA22
Genomic Map