siRNA Details
| Gene Symbol: MYO18B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318245.1 | NP_001305174.1 | 23 | 4265 | ||
| NM_032608.6 | NP_115997.5 | 23 | 4262 | |||
| XM_011530458.2 | XP_011528760.1 | 24 | 4401 | |||
| XM_011530459.2 | XP_011528761.1 | 24 | 4372 | |||
| XM_011530460.2 | XP_011528762.1 | 23 | 4245 | |||
| XM_011530461.2 | XP_011528763.1 | 24 | 4401 | |||
| XM_011530464.2 | XP_011528766.1 | 24 | 4398 | |||
| XM_011530465.2 | XP_011528767.1 | 23 | 4158 | |||
| XM_011530466.2 | XP_011528768.1 | 24 | 4402 | |||
| XM_017029012.1 | XP_016884501.1 | 24 | 4401 | |||
| XM_017029013.1 | XP_016884502.1 | 24 | 4449 | |||
| XM_017029014.1 | XP_016884503.1 | 24 | 4401 | |||
| XM_017029015.1 | XP_016884504.1 | 23 | 4272 | |||
| XM_017029016.1 | XP_016884505.1 | 12 | 1525 | |||
| XM_017029017.1 | XP_016884506.1 | 22 | 4126 | |||
| GenBank mRNA | AB042648.2 | - | 19 | 2574 | ||
| AB075376.1 | - | 23 | 4262 | |||
| AJ310931.2 | - | 23 | 4253 | |||
| AY077700.1 | - | 23 | 4175 | |||
| BC144596.1 | - | 21 | 3917 | |||
| BC144597.1 | - | 22 | 4062 | 9053127 | ||
| BC150626.1 | - | 22 | 4062 | 9056996 | ||

MYO18B
Genomic Map