siRNA Details
| Gene Symbol: PLEKHA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032639.3 | NP_116028.1 | 13 | 2005 | ||
| XM_011515592.2 | XP_011513894.1 | 14 | 2015 | |||
| GenBank mRNA | BC002838.2 | - | 13 | 1694 | 3635744 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLEKHA8| Gene Symbol: PLEKHA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032639.3 | NP_116028.1 | 13 | 2005 | ||
| XM_011515592.2 | XP_011513894.1 | 14 | 2015 | |||
| GenBank mRNA | BC002838.2 | - | 13 | 1694 | 3635744 | |