siRNA Details
| Gene Symbol: USP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032663.4 | NP_116052.2 | 1, 2 | 200 | ||
| XM_005253962.3 | XP_005254019.1 | 1, 2 | 186 | |||
| XM_005253965.4 | XP_005254022.1 | 1, 2 | 194 | |||
| XM_017020048.1 | XP_016875537.1 | 1, 2 | 195 | |||
| XM_017020049.1 | XP_016875538.1 | 1, 2 | 186 | |||
| XM_017020053.1 | XP_016875542.1 | 1, 2 | 110 | |||
| GenBank mRNA | AJ586136.1 | - | 1, 2 | 91 | ||
| AK027820.1 | - | 1, 2 | 165 | |||
| BC004868.2 | - | 1, 2 | 190 | 3833242 | ||

USP30
Genomic Map