siRNA Details
Gene Symbol: B3GNT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033309.2 | NP_171608.2 | 2 | 388 | ||
GenBank mRNA | AY358485.1 | - | 2 | 320 | ||
CF994177.1 | - | 2 | 415 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B3GNT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033309.2 | NP_171608.2 | 2 | 388 | ||
GenBank mRNA | AY358485.1 | - | 2 | 320 | ||
CF994177.1 | - | 2 | 415 |