siRNA Details
| Gene Symbol: BUD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159736.1 | NP_001153208.1 | 8 | 1238 | ||
| NM_032725.3 | NP_116114.1 | 8 | 1640 | |||
| XM_011543035.2 | XP_011541337.1 | 8 | 2214 | |||
| GenBank mRNA | AK057832.1 | - | 8 | 1227 | ||
| AK130600.1 | - | 1 | 1124 | |||
| AK225164.1 | - | 8 | 1227 | |||
| AK289635.1 | - | 8 | 1629 | |||
| AK291111.1 | - | 8 | 1629 | |||
| BC006350.2 | - | 8 | 1630 | 4111572 | ||

BUD13
Genomic Map