siRNA Details
| Gene Symbol: CCNI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006835.2 | NP_006826.1 | 3, 4 | 788 | ||
| GenBank mRNA | AB209766.1 | - | 3, 4 | 790 | ||
| AF135162.1 | - | 3, 4 | 443 | |||
| AK222931.1 | - | 2, 3 | 240 | |||
| AK312632.1 | - | 5, 6 | 658 | |||
| BC000420.2 | - | 3, 4 | 787 | 2964432 | ||
| BC004975.1 | - | 3, 4 | 787 | 2957878 | ||
| BT020019.1 | - | 2, 3 | 244 | |||
| CR541783.1 | - | 2, 3 | 244 | |||
| D50310.1 | - | 2, 3 | 244 | |||

CCNI
Genomic Map