siRNA Details
| Gene Symbol: TRIM52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032765.2 | NP_116154.1 | 1 | 987 | ||
| XM_005266000.3 | XP_005266057.1 | 1 | 1029 | |||
| XM_017009991.1 | XP_016865480.1 | 1 | 1029 | |||
| XM_017009992.1 | XP_016865481.1 | 1 | 1029 | |||
| XM_017009993.1 | XP_016865482.1 | 1 | 1029 | |||
| XM_017009994.1 | XP_016865483.1 | 1 | 1029 | |||
| GenBank mRNA | AB209243.1 | - | 1 | 1005 | ||
| AK054802.1 | - | 1 | 964 | |||
| BC007372.2 | - | 1 | 987 | 3636274 | ||
| JX896146.1 | - | 1 | 682 | |||

TRIM52
Genomic Map