siRNA Details
| Gene Symbol: CCDC142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032779.3 | NP_116168.3 | 6, 7 | 1982 | ||
| GenBank mRNA | AK027303.1 | - | 1, 2 | 67 | ||
| AK075543.1 | - | 6, 7 | 1748 | |||
| BC117269.1 | - | 6, 7 | 1700 | 40125820 | ||
| BC143398.1 | - | 6, 7 | 1700 | 9051905 | ||
| BC143399.1 | - | 6, 7 | 1700 | |||

CCDC142
Genomic Map