siRNA Details
| Gene Symbol: CCDC142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032779.3 | NP_116168.3 | 1 | 728 | ||
| GenBank mRNA | AK075543.1 | - | 1 | 494 | ||
| BC117269.1 | - | 1 | 425 | 40125820 | ||
| BC143398.1 | - | 1 | 425 | 9051905 | ||
| BC143399.1 | - | 1 | 425 | |||

CCDC142
Genomic Map