siRNA Details
| Gene Symbol: ZBTB45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316978.1 | NP_001303907.1 | 2 | 1401 | ||
| NM_001316979.1 | NP_001303908.1 | 2 | 1128 | |||
| NM_001316980.1 | NP_001303909.1 | 2 | 951 | |||
| NM_001316981.1 | NP_001303910.1 | 2 | 1253 | |||
| NM_001316982.1 | NP_001303911.1 | 2 | 1071 | |||
| NM_032792.3 | NP_116181.1 | 2 | 1157 | |||
| XM_006723445.3 | XP_006723508.1 | 2 | 1171 | |||
| GenBank mRNA | AK027392.1 | - | 2 | 940 | ||
| AK130056.1 | - | 3 | 758 | |||
| BC024738.1 | - | 2 | 1152 | 5110354 | ||

ZBTB45
Genomic Map